Product Details

SNP ID
rs1007409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41184759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACACAGCTGGGGCAGCAACAGGT[C/G]GTCTGGCAGCAGCTGGGGCGGCAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP4-1 PubMed Links

Gene Details

Gene
KRTAP4-1
Gene Name
keratin associated protein 4-1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033060.2 137 Silent Mutation ACC,ACG T32T NP_149049.1
Gene
KRTAP4-2
Gene Name
keratin associated protein 4-2
There are no transcripts associated with this gene.

Gene
KRTAP9-1
Gene Name
keratin associated protein 9-1
There are no transcripts associated with this gene.

View Full Product Details