Product Details
- SNP ID
-
rs11391
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:113940851 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AATTTTAAATACTATCACATAAAGC[A/C]TGGTGGAGAAAAGAAATTTTGCTTC
- Phenotype
-
MIM: 605164
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HDAC2
PubMed Links
Gene Details
- Gene
- HDAC2
- Gene Name
- histone deacetylase 2
View Full Product Details