Product Details

SNP ID
rs376790302
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75762731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAGCTCTTCCAGTTGTACCTCCC[A/G]GAGGCTTTCCTTGCCCAGCGCCCGG
Phenotype
MIM: 604313 MIM: 147557
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALK1 PubMed Links

Gene Details

Gene
GALK1
Gene Name
galactokinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000154.1 829 Missense Mutation CGG,TGG R256W NP_000145.1
Gene
ITGB4
Gene Name
integrin subunit beta 4
There are no transcripts associated with this gene.

View Full Product Details