Product Details

SNP ID
rs2035549
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:185589204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGGGAAGAATGGAACGTCTCTC[C/T]GCTTCCACCACAAACTAAAGTTAAG
Phenotype
MIM: 616349
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SORBS2 PubMed Links

Gene Details

Gene
SORBS2
Gene Name
sorbin and SH3 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145670.1 Intron NP_001139142.1
NM_001145671.2 Intron NP_001139143.1
NM_001145672.1 Intron NP_001139144.1
NM_001145673.1 Intron NP_001139145.1
NM_001145674.1 Intron NP_001139146.1
NM_001145675.1 Intron NP_001139147.1
NM_001270771.1 Intron NP_001257700.1
NM_003603.6 Intron NP_003594.3
NM_021069.4 Intron NP_066547.1
XM_005263307.1 Intron XP_005263364.1
XM_005263308.1 Intron XP_005263365.1
XM_005263310.1 Intron XP_005263367.1
XM_005263311.1 Intron XP_005263368.1
XM_005263312.1 Intron XP_005263369.1
XM_005263313.1 Intron XP_005263370.1
XM_006714388.1 Intron XP_006714451.1
XM_006714390.1 Intron XP_006714453.1
XM_017008738.1 Intron XP_016864227.1
XM_017008739.1 Intron XP_016864228.1
XM_017008740.1 Intron XP_016864229.1
XM_017008741.1 Intron XP_016864230.1
XM_017008742.1 Intron XP_016864231.1
XM_017008743.1 Intron XP_016864232.1
XM_017008744.1 Intron XP_016864233.1
XM_017008745.1 Intron XP_016864234.1
XM_017008746.1 Intron XP_016864235.1
XM_017008747.1 Intron XP_016864236.1
XM_017008748.1 Intron XP_016864237.1
XM_017008749.1 Intron XP_016864238.1
XM_017008750.1 Intron XP_016864239.1
XM_017008751.1 Intron XP_016864240.1
XM_017008752.1 Intron XP_016864241.1
XM_017008753.1 Intron XP_016864242.1
XM_017008754.1 Intron XP_016864243.1
XM_017008755.1 Intron XP_016864244.1
XM_017008756.1 Intron XP_016864245.1
XM_017008757.1 Intron XP_016864246.1
XM_017008758.1 Intron XP_016864247.1
XM_017008759.1 Intron XP_016864248.1
XM_017008760.1 Intron XP_016864249.1
XM_017008761.1 Intron XP_016864250.1
XM_017008762.1 Intron XP_016864251.1
XM_017008763.1 Intron XP_016864252.1
XM_017008764.1 Intron XP_016864253.1
XM_017008765.1 Intron XP_016864254.1
XM_017008766.1 Intron XP_016864255.1
XM_017008767.1 Intron XP_016864256.1
XM_017008768.1 Intron XP_016864257.1
XM_017008769.1 Intron XP_016864258.1
XM_017008770.1 Intron XP_016864259.1
XM_017008771.1 Intron XP_016864260.1
XM_017008772.1 Intron XP_016864261.1
XM_017008773.1 Intron XP_016864262.1
XM_017008774.1 Intron XP_016864263.1
XM_017008775.1 Intron XP_016864264.1

View Full Product Details