Product Details

SNP ID
rs1687228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184231061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCATCCTGGTATTTAGCAGTCG[C/T]GGGCTGGGCACCTGACCTGCGCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VWA5B2 PubMed Links

Gene Details

Gene
VWA5B2
Gene Name
von Willebrand factor A domain containing 5B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320373.1 Intron NP_001307302.1
NM_138345.2 Intron NP_612354.1
XM_011513288.2 Intron XP_011511590.1
XM_011513289.2 Intron XP_011511591.1
XM_011513290.2 Intron XP_011511592.1
XM_011513291.2 Intron XP_011511593.1
XM_011513292.2 Intron XP_011511594.1

View Full Product Details