Product Details
- SNP ID
-
rs34606895
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:122660798 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGGTGGCTAGAACTTTGTCTAC[A/C]ACAAATGGCTTTTTAATGTTTCCAG
- Phenotype
-
MIM: 609201
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
UBASH3B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143366067] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UBASH3B
- Gene Name
- ubiquitin associated and SH3 domain containing B
View Full Product Details