Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001388.4 | 286 | Silent Mutation | TCA,TCG | S49S | NP_001379.1 |
XM_005256499.2 | 286 | Silent Mutation | TCA,TCG | S49S | XP_005256556.1 |
XM_005256500.2 | 286 | UTR 5 | XP_005256557.1 | ||
XM_011523704.1 | 286 | Silent Mutation | TCA,TCG | S49S | XP_011522006.1 |
XM_011523705.1 | 286 | Silent Mutation | TCA,TCG | S49S | XP_011522007.1 |
XM_011523706.1 | 286 | UTR 5 | XP_011522008.1 | ||
XM_011523707.1 | 286 | UTR 5 | XP_011522009.1 | ||
XM_017024296.1 | 286 | UTR 5 | XP_016879785.1 | ||
XM_017024297.1 | 286 | UTR 5 | XP_016879786.1 |