Product Details

SNP ID
rs3008633
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:222671498 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACATACATTTAATACTGCCGCTGA[C/T]GACAAAACTCACAAGTCACTAGAAG
Phenotype
MIM: 612375 MIM: 613455
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIDA PubMed Links

Gene Details

Gene
AIDA
Gene Name
axin interactor, dorsalization associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022831.2 Intron NP_073742.2
Gene
MIA3
Gene Name
MIA family member 3, ER export factor
There are no transcripts associated with this gene.

View Full Product Details