Product Details

SNP ID
rs2302181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2477817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAGATCCAGGAGCTGGCGGCCGC[C/T]GATCCGATGGAGAAGGGGGGACCCA
Phenotype
MIM: 604948
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GADD45B PubMed Links

Gene Details

Gene
GADD45B
Gene Name
growth arrest and DNA damage inducible beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015675.3 933 UTR 3 NP_056490.2
XM_017026822.1 933 UTR 3 XP_016882311.1

View Full Product Details