Product Details

SNP ID
rs2303094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33209375 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCAGAAAGAAAATGGGCACCCC[A/C]GTAAGGATGATGATGACGCCGACCC
Phenotype
MIM: 603159 MIM: 607959
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LRP3 PubMed Links

Gene Details

Gene
LRP3
Gene Name
LDL receptor related protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002333.3 1156 Intron NP_002324.2
XM_005258945.1 1156 Intron XP_005259002.1
Gene
SLC7A10
Gene Name
solute carrier family 7 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019849.2 1156 Silent Mutation ACA,ACC T458T NP_062823.1
XM_006723284.2 1156 Silent Mutation ACA,ACC T379T XP_006723347.1
XM_011527119.1 1156 Silent Mutation ACA,ACC T595T XP_011525421.1
XM_011527120.1 1156 Silent Mutation ACA,ACC T568T XP_011525422.1

View Full Product Details