Product Details

SNP ID
rs3027470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49034200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTGCTGACAGTGATGGCTGGTG[T/C]GGCCACGCCTTGACTACTGTACACA
Phenotype
MIM: 314310
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
TFE3 PubMed Links

Gene Details

Gene
TFE3
Gene Name
transcription factor binding to IGHM enhancer 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282142.1 1228 Missense Mutation ACA,GCA T208A NP_001269071.1
NM_006521.5 1228 Missense Mutation ACA,GCA T313A NP_006512.2

View Full Product Details