Product Details

SNP ID
rs2385074
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.8:123042466 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGTGGGAAAACAGTGGTTGCCTGGA[A/T]CAACGGCAAACGCTCCGGCCAGTGG
Phenotype
MIM: 608813
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DERL1 PubMed Links

Gene Details

Gene
DERL1
Gene Name
derlin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134671.2 Intron NP_001128143.1
NM_024295.5 Intron NP_077271.1
XM_006716657.1 Intron XP_006716720.1
XM_011517299.1 Intron XP_011515601.1
Gene
TBC1D31
Gene Name
TBC1 domain family member 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145088.1 Intron NP_001138560.1
NM_145647.3 Intron NP_663622.2
XM_005251102.1 Intron XP_005251159.1
XM_005251103.1 Intron XP_005251160.1
XM_005251104.1 Intron XP_005251161.1
XM_011517378.2 Intron XP_011515680.1
XM_011517379.2 Intron XP_011515681.1
XM_017013983.1 Intron XP_016869472.1
XM_017013984.1 Intron XP_016869473.1
XM_017013985.1 Intron XP_016869474.1
XM_017013986.1 Intron XP_016869475.1
XM_017013987.1 Intron XP_016869476.1
XM_017013988.1 Intron XP_016869477.1
XM_017013989.1 Intron XP_016869478.1
XM_017013990.1 Intron XP_016869479.1

View Full Product Details