Product Details

SNP ID
rs2142719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32477194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGAATAGCTTCTATTTCAGTAA[C/T]TTTAATTTCTCTACCAAGGACACTT
Phenotype
MIM: 605648
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO7 PubMed Links

Gene Details

Gene
FBXO7
Gene Name
F-box protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033024.1 Intron NP_001028196.1
NM_001257990.1 Intron NP_001244919.1
NM_012179.3 Intron NP_036311.3
XM_011530106.1 Intron XP_011528408.1

View Full Product Details