Product Details

SNP ID
rs2231423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73234216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTCAGGTCCTTCTAGCCGCAC[C/G]GAGTCGGTGGTAACGAACCATAGAG
Phenotype
MIM: 605140
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCT7 PubMed Links

Gene Details

Gene
CCT7
Gene Name
chaperonin containing TCP1 subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009570.2 Intron NP_001009570.1
NM_001166284.1 Intron NP_001159756.1
NM_001166285.1 Intron NP_001159757.1
NM_006429.3 Intron NP_006420.1
XM_011532478.2 Intron XP_011530780.1
XM_011532479.1 Intron XP_011530781.1
Gene
PRADC1
Gene Name
protease associated domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032319.1 Intron NP_115695.1
Gene
SMYD5
Gene Name
SMYD family member 5
There are no transcripts associated with this gene.

View Full Product Details