Product Details

SNP ID
rs2232145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:61131754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTTCCAGTTGACCCTCGAATCTC[A/G]GCGATGGCTGGGCCAAAGGTTGAGC
Phenotype
MIM: 153340 MIM: 610038
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD5 PubMed Links

Gene Details

Gene
CD5
Gene Name
CD5 molecule
There are no transcripts associated with this gene.

Gene
VPS37C
Gene Name
VPS37C, ESCRT-I subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017966.4 1235 UTR 3 NP_060436.4
XM_005274077.3 1235 UTR 3 XP_005274134.1
XM_017017955.1 1235 UTR 3 XP_016873444.1

View Full Product Details