Product Details
- SNP ID
-
rs2069714
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:68157264 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGCAACCATTCTTGCTTCTAATTGG[G/T]CAGTACAATCTGATAGGTTGGCTAG
- Phenotype
-
MIM: 147570
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
IFNG
PubMed Links
Gene Details
- Gene
- IFNG
- Gene Name
- interferon gamma
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000619.2 |
|
Intron |
|
|
NP_000610.2 |
View Full Product Details