Product Details
- SNP ID
-
rs2070032
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:154584990 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATAGGGAAAGGATGCACTTACAGT[A/G]GGATAGAACTTATGCAAATAAGCGG
- Phenotype
-
MIM: 134820
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGA
PubMed Links
Gene Details
- Gene
- FGA
- Gene Name
- fibrinogen alpha chain
View Full Product Details