Product Details

SNP ID
rs2241547
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:77977206 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAGTCCAGGGAAGCAGTGTGTGCA[C/G]GAGCTGTGGGGCCGTCTATAGATTA
Phenotype
MIM: 614258
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
POLR3A PubMed Links

Gene Details

Gene
POLR3A
Gene Name
RNA polymerase III subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007055.3 4578 UTR 3 NP_008986.2

View Full Product Details