Product Details

SNP ID
rs2281703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24082358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGGACCTTCTCCCCTTCTCCTGCC[C/T]TCTGTCCCCTGTCCCAGTCCATAAC
Phenotype
MIM: 605688 MIM: 162080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPNE6 PubMed Links

Gene Details

Gene
CPNE6
Gene Name
copine 6
There are no transcripts associated with this gene.

Gene
NRL
Gene Name
neural retina leucine zipper
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006177.3 Intron NP_006168.1
XM_005267708.4 Intron XP_005267765.1
XM_005267709.3 Intron XP_005267766.1
XM_005267710.3 Intron XP_005267767.1
XM_011536801.2 Intron XP_011535103.2
XM_011536802.1 Intron XP_011535104.1
XM_011536804.2 Intron XP_011535106.1
XM_011536805.2 Intron XP_011535107.1
XM_011536806.2 Intron XP_011535108.2
XM_017021350.1 Intron XP_016876839.1

View Full Product Details