Product Details

SNP ID
rs2269899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112944151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGAGAGAGTTGCCAGGCTGCAGG[C/T]ACTGCTGACTCAGCTGGAAGCAACT
Phenotype
MIM: 603351
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OAS3 PubMed Links

Gene Details

Gene
OAS3
Gene Name
2'-5'-oligoadenylate synthetase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006187.3 Intron NP_006178.2
XM_005253889.3 Intron XP_005253946.1
XM_017019363.1 Intron XP_016874852.1

View Full Product Details