Product Details

SNP ID
rs2270003
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:54783029 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GAAGAATGGCAGGTGGCCCCAACTG[A/C]CAAGCATCACATTACAAATGATCTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TTC22 PubMed Links

Gene Details

Gene
TTC22
Gene Name
tetratricopeptide repeat domain 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114108.1 Intron NP_001107580.1
NM_017904.3 Intron NP_060374.2
XM_011541671.2 Intron XP_011539973.1
XM_017001582.1 Intron XP_016857071.1

View Full Product Details