Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130831.1 | 6130 | UTR 3 | NP_001124303.1 | ||
NM_003644.2 | 6130 | UTR 3 | NP_003635.2 | ||
NM_201432.1 | 6130 | UTR 3 | NP_958836.1 | ||
NM_201433.1 | 6130 | UTR 3 | NP_958839.1 | ||
XM_005256831.3 | 6130 | UTR 3 | XP_005256888.1 | ||
XM_005256832.3 | 6130 | UTR 3 | XP_005256889.1 | ||
XM_005256833.3 | 6130 | UTR 3 | XP_005256890.1 | ||
XM_005256834.3 | 6130 | UTR 3 | XP_005256891.1 | ||
XM_005256835.3 | 6130 | UTR 3 | XP_005256892.1 | ||
XM_011524044.2 | 6130 | UTR 3 | XP_011522346.1 | ||
XM_017025239.1 | 6130 | UTR 3 | XP_016880728.1 | ||
XM_017025240.1 | 6130 | UTR 3 | XP_016880729.1 | ||
XM_017025241.1 | 6130 | UTR 3 | XP_016880730.1 | ||
XM_017025242.1 | 6130 | UTR 3 | XP_016880731.1 |