Product Details

SNP ID
rs2534699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6557151 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTTTCTCAAAAGGCTGCTCTG[C/G]CAACTGCTGCTTCTCAACCCCCTTG
Phenotype
MIM: 610495 MIM: 164031
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IFFO1 PubMed Links

Gene Details

Gene
IFFO1
Gene Name
intermediate filament family orphan 1
There are no transcripts associated with this gene.

Gene
NOP2
Gene Name
NOP2 nucleolar protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033714.2 2399 Missense Mutation CCA,GCA P757A NP_001028886.1
NM_001258308.1 2399 Missense Mutation CCA,GCA P761A NP_001245237.1
NM_001258309.1 2399 Missense Mutation CCA,GCA P794A NP_001245238.1
NM_001258310.1 2399 UTR 3 NP_001245239.1
NM_006170.3 2399 Missense Mutation CCA,GCA P757A NP_006161.2
XM_005253691.1 2399 Missense Mutation CCA,GCA P761A XP_005253748.1

View Full Product Details