Product Details

SNP ID
rs2545762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40800481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCAGGCCCGGTGGCCCCCAGCTG[C/T]ATCAAGTGGAGGCGGAGGAGGAGGC
Phenotype
MIM: 606424 MIM: 612945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP2T1P PubMed Links

Gene Details

Gene
CYP2T1P
Gene Name
cytochrome P450 family 2 subfamily T member 1, pseudogene
There are no transcripts associated with this gene.

Gene
EGLN2
Gene Name
egl-9 family hypoxia inducible factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053046.3 263 UTR 5 NP_444274.1
NM_080732.3 263 UTR 5 NP_542770.2
Gene
MIA-RAB4B
Gene Name
MIA-RAB4B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
RAB4B
Gene Name
RAB4B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
RAB4B-EGLN2
Gene Name
RAB4B-EGLN2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details