Product Details
- SNP ID
-
rs2617691
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53476035 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTGCCTCTAAAAATTCCTATTTGA[C/T]GTTGGGTGGCCGGGGTTGGAAGGGC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF813
PubMed Links
Gene Details
- Gene
- ZNF813
- Gene Name
- zinc finger protein 813
View Full Product Details