Product Details
- SNP ID
-
rs2598047
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:37848886 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCTGGAGAAGAAAGCCCAGCAGGA[G/T]GCCTGCTAGCCTCAGCCTCGGGCCC
- Phenotype
-
MIM: 607421
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NME8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115132982] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NME8
- Gene Name
- NME/NM23 family member 8
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016616.4 |
195 |
UTR 5 |
|
|
NP_057700.3 |
View Full Product Details