Product Details
- SNP ID
-
rs2642423
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:220788453 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGACATAGTTGAAAGGTGAGTGACG[C/T]ATGCATCTTTGCTTCAGGAAGGAAG
- Phenotype
-
MIM: 614126
MIM: 614127
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MARC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117574394] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MARC1
- Gene Name
- mitochondrial amidoxime reducing component 1
- Gene
- MARC2
- Gene Name
- mitochondrial amidoxime reducing component 2
There are no transcripts associated with this gene.
View Full Product Details