Product Details

SNP ID
rs2263696
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18293937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAGAATGAAAGGGGGAAAAAGAAC[A/T]TTGTCATTTGGTAAACAGCACAGTA
Phenotype
MIM: 604075
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF133 PubMed Links

Gene Details

Gene
ZNF133
Gene Name
zinc finger protein 133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083330.2 4120 Intron NP_001076799.1
NM_001282995.1 4120 Intron NP_001269924.1
NM_001282996.1 4120 Intron NP_001269925.1
NM_001282997.1 4120 Intron NP_001269926.1
NM_001282998.1 4120 Intron NP_001269927.1
NM_001282999.1 4120 Intron NP_001269928.1
NM_001283000.1 4120 Intron NP_001269929.1
NM_001283001.1 4120 Intron NP_001269930.1
NM_001283002.1 4120 Intron NP_001269931.1
NM_001283003.1 4120 Intron NP_001269932.1
NM_001283004.1 4120 Intron NP_001269933.1
NM_001283005.1 4120 Intron NP_001269934.1
NM_001283006.1 4120 Intron NP_001269935.1
NM_001283007.1 4120 Intron NP_001269936.1
NM_001283008.1 4120 Intron NP_001269937.1
NM_003434.5 4120 Intron NP_003425.2
XM_005260819.1 4120 Intron XP_005260876.1
XM_005260820.4 4120 Intron XP_005260877.1
XM_011529336.1 4120 Intron XP_011527638.1
XM_011529337.1 4120 Intron XP_011527639.1
XM_011529338.2 4120 Intron XP_011527640.1
XM_011529339.1 4120 Intron XP_011527641.1
XM_011529340.1 4120 Intron XP_011527642.1
XM_017028041.1 4120 UTR 5 XP_016883530.1
XM_017028042.1 4120 Intron XP_016883531.1
XM_017028043.1 4120 UTR 5 XP_016883532.1
XM_017028044.1 4120 UTR 5 XP_016883533.1
XM_017028045.1 4120 UTR 5 XP_016883534.1
XM_017028046.1 4120 UTR 5 XP_016883535.1
XM_017028047.1 4120 Intron XP_016883536.1
XM_017028048.1 4120 Intron XP_016883537.1
XM_017028049.1 4120 UTR 5 XP_016883538.1
XM_017028050.1 4120 Intron XP_016883539.1
XM_017028051.1 4120 Intron XP_016883540.1
XM_017028052.1 4120 Intron XP_016883541.1
XM_017028053.1 4120 Intron XP_016883542.1
XM_017028054.1 4120 Intron XP_016883543.1
XM_017028055.1 4120 Intron XP_016883544.1
XM_017028056.1 4120 Intron XP_016883545.1
XM_017028057.1 4120 Intron XP_016883546.1
XM_017028058.1 4120 Intron XP_016883547.1

View Full Product Details