Product Details

SNP ID
rs2908275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:44144659 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAAGAAAATGGCCAGGACTGGGT[A/G]ATGAGCAGCCCCTGGCTGCCCTTTT
Phenotype
MIM: 138079 MIM: 613993
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCK PubMed Links

Gene Details

Gene
GCK
Gene Name
glucokinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000162.3 2345 UTR 3 NP_000153.1
NM_033507.1 2345 UTR 3 NP_277042.1
NM_033508.1 2345 UTR 3 NP_277043.1
XM_017011966.1 2345 Intron XP_016867455.1
Gene
MYL7
Gene Name
myosin light chain 7
There are no transcripts associated with this gene.

View Full Product Details