Product Details

SNP ID
rs2234234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:9629358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGACAGGATAGACAGCAACGC[A/G]CTGATGGGTGCACCCCTGCCAGGAA
Phenotype
MIM: 604796
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS2R1 PubMed Links

Gene Details

Gene
TAS2R1
Gene Name
taste 2 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019599.2 994 Silent Mutation AGC,AGT S225S NP_062545.1

View Full Product Details