Product Details

SNP ID
rs2270243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17085614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGGGGCTGTGGAGAAGGGGCC[C/T]GGGGAGGAGCCAGGACGGGTCTGGA
Phenotype
MIM: 605461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL17RA PubMed Links

Gene Details

Gene
IL17RA
Gene Name
interleukin 17 receptor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289905.1 Intron NP_001276834.1
NM_014339.6 Intron NP_055154.3

View Full Product Details