Product Details

SNP ID
rs1805808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:90006832 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATTAGAGCTACTGGGGACTAAAA[C/T]GAACTGGGCTTGGTTTGAATGTATG
Phenotype
MIM: 222745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DECR1 PubMed Links

Gene Details

Gene
DECR1
Gene Name
2,4-dienoyl-CoA reductase 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001359.1 Intron NP_001350.1
XM_005250808.4 Intron XP_005250865.1
XM_011516880.2 Intron XP_011515182.1
XM_017013147.1 Intron XP_016868636.1
XM_017013148.1 Intron XP_016868637.1

View Full Product Details