Product Details

SNP ID
rs2975307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144879096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAACTAACACCTTAGTTATTTTAT[C/T]TATCTGAAACAACACCTCACAGAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF250 PubMed Links

Gene Details

Gene
ZNF250
Gene Name
zinc finger protein 250
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109689.3 4219 UTR 3 NP_001103159.1
NM_021061.4 4219 UTR 3 NP_066405.1
XM_005272327.1 4219 Intron XP_005272384.1
XM_005272328.3 4219 Intron XP_005272385.1
XM_006716612.3 4219 UTR 3 XP_006716675.1
XM_006716613.3 4219 UTR 3 XP_006716676.1
XM_011517209.1 4219 Intron XP_011515511.1

View Full Product Details