Product Details

SNP ID
rs2278225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:78487278 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATTCCATCTCTGTATAATTATCT[A/G]TTCTGCACCAAGATACGGTGCTTTA
Phenotype
MIM: 609718 MIM: 606911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LHFPL2 PubMed Links

Gene Details

Gene
LHFPL2
Gene Name
lipoma HMGIC fusion partner-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005779.2 2982 UTR 3 NP_005770.1
XM_006714515.1 2982 UTR 3 XP_006714578.1
Gene
SCAMP1
Gene Name
secretory carrier membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details