Product Details

SNP ID
rs2292974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27460874 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCAGCAAGGAAGGAGAGGAATGA[C/T]GTCAAGGAGAAGCCTGGAGGCTCAA
Phenotype
MIM: 118502 MIM: 601212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNA2 PubMed Links

Gene Details

Gene
CHRNA2
Gene Name
cholinergic receptor nicotinic alpha 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000742.3 2818 UTR 3 NP_000733.2
NM_001282455.1 2818 UTR 3 NP_001269384.1
XM_005273397.2 2818 UTR 3 XP_005273454.1
XM_006716282.1 2818 UTR 3 XP_006716345.1
XM_011544388.1 2818 UTR 3 XP_011542690.1
XM_011544389.1 2818 UTR 3 XP_011542691.1
XM_017013003.1 2818 UTR 3 XP_016868492.1
XM_017013004.1 2818 UTR 3 XP_016868493.1
Gene
PTK2B
Gene Name
protein tyrosine kinase 2 beta
There are no transcripts associated with this gene.

View Full Product Details