Product Details

SNP ID
rs2305142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:97759259 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTGCACGAGCTGATATGCCACC[A/G]ATGACATCCAAATGCAAACTTTCCC
Phenotype
MIM: 615659
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM131 PubMed Links

Gene Details

Gene
TMEM131
Gene Name
transmembrane protein 131
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015348.1 Intron NP_056163.1
XM_005263909.1 Intron XP_005263966.1
XM_005263910.1 Intron XP_005263967.1
XM_005263911.1 Intron XP_005263968.1
XM_005263912.3 Intron XP_005263969.1
XM_011510869.1 Intron XP_011509171.1

View Full Product Details