Product Details

SNP ID
rs3020099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:92257235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAGAAAAAAAAGAAAAGAAAA[A/T]TTGTTACAATGAAGAAGTCGTTATG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ANKIB1 PubMed Links

Gene Details

Gene
ANKIB1
Gene Name
ankyrin repeat and IBR domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019004.1 Intron NP_061877.1
XM_005250458.3 Intron XP_005250515.1
XM_011516334.1 Intron XP_011514636.1
XM_011516335.2 Intron XP_011514637.1
XM_017012359.1 Intron XP_016867848.1
XM_017012360.1 Intron XP_016867849.1

View Full Product Details