Product Details

SNP ID
rs2353603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:217759 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTAGGGGGAGGAATATCAGCATTAA[C/T]AGGAGACTTGCTATAAGAAGCTATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107986244 PubMed Links

Gene Details

Gene
LOC107986244
Gene Name
zinc finger protein 141-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017008889.1 Intron XP_016864378.1
Gene
ZNF876P
Gene Name
zinc finger protein 876, pseudogene
There are no transcripts associated with this gene.

View Full Product Details