Product Details

SNP ID
rs2453436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86869646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTCTTCACAGTTTGCTAACCTTA[A/G]GGGAGGAATGAATGTTCTCAAAAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNBD1 PubMed Links

Gene Details

Gene
CNBD1
Gene Name
cyclic nucleotide binding domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173538.2 Intron NP_775809.1
XM_017013149.1 Intron XP_016868638.1

View Full Product Details