Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_015533.3 | Intron | NP_056348.2 | |||
XM_011544912.2 | Intron | XP_011543214.1 | |||
XM_017017517.1 | Intron | XP_016873006.1 | |||
XM_017017518.1 | Intron | XP_016873007.1 | |||
XM_017017519.1 | Intron | XP_016873008.1 | |||
XM_017017520.1 | Intron | XP_016873009.1 | |||
XM_017017521.1 | Intron | XP_016873010.1 | |||
XM_017017522.1 | Intron | XP_016873011.1 | |||
XM_017017523.1 | Intron | XP_016873012.1 | |||
XM_017017524.1 | Intron | XP_016873013.1 |