Product Details

SNP ID
rs2540450
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.2:201627272 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAAAAAGAATGGAAGGATAAGAAAA[A/G]TTGCTGTTATTGCCATTAACAATTG
Phenotype
MIM: 614423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALS2CR11 PubMed Links

Gene Details

Gene
ALS2CR11
Gene Name
amyotrophic lateral sclerosis 2 chromosome region candidate 11
There are no transcripts associated with this gene.

Gene
TMEM237
Gene Name
transmembrane protein 237
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001044385.2 Intron NP_001037850.1
NM_152388.3 Intron NP_689601.2

View Full Product Details