Product Details

SNP ID
rs2525990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56884299 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGGGAGAAGGAGATGGGAGCCAG[A/G]CACTCATCCCAATCCCAGAGACTCC
Phenotype
MIM: 600453
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR3614 PubMed Links

Gene Details

Gene
MIR3614
Gene Name
microRNA 3614
There are no transcripts associated with this gene.

Gene
MTVR2
Gene Name
mouse mammary tumor virus receptor homolog 2
There are no transcripts associated with this gene.

Gene
TRIM25
Gene Name
tripartite motif containing 25
There are no transcripts associated with this gene.

View Full Product Details