Product Details
- SNP ID
-
rs2568085
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:8954201 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATACATGAAATGTCCATGTATGCAG[A/C]GACAGAAAGTAGACTAGTGACCGAG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM9B
PubMed Links
Gene Details
- Gene
- TMEM9B
- Gene Name
- TMEM9 domain family member B
View Full Product Details