Product Details
- SNP ID
-
rs2767401
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:127868565 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCATTCCTGCCCCCTAGGGCCAT[C/G]TCCTCCCCATCTTCCCATCTATGAA
- Phenotype
-
MIM: 103000
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AK1
PubMed Links
Gene Details
- Gene
- AK1
- Gene Name
- adenylate kinase 1
- Gene
- MIR4672
- Gene Name
- microRNA 4672
There are no transcripts associated with this gene.
View Full Product Details