Product Details

SNP ID
rs2129580
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45722491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGAGACCTCAAAGGTATGAAG[C/T]TAAGGCAGAAAATAAATCCAAAGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPINT4 PubMed Links

Gene Details

Gene
SPINT4
Gene Name
serine peptidase inhibitor, Kunitz type 4
There are no transcripts associated with this gene.

Gene
WFDC3
Gene Name
WAP four-disulfide core domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080614.1 Intron NP_542181.1

View Full Product Details