Product Details
- SNP ID
-
rs2145542
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:22949827 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTATCTTGCTGAATAAAAAGAGTC[C/T]TTACAAAGGTCCTTGTTTTGGCCGG
- Phenotype
-
MIM: 613431
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HAUS4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8020125] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HAUS4
- Gene Name
- HAUS augmin like complex subunit 4
- Gene
- LOC101926933
- Gene Name
- uncharacterized LOC101926933
There are no transcripts associated with this gene.
- Gene
- MIR4707
- Gene Name
- microRNA 4707
There are no transcripts associated with this gene.
View Full Product Details