Product Details

SNP ID
rs2879515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:34579095 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCACAGGCAAGAGGCCTCATGCGC[A/G]CATTGCAGATGCTGAGAAAGACTCT
Phenotype
MIM: 609619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLGA8B PubMed Links

Gene Details

Gene
GOLGA8B
Gene Name
golgin A8 family member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023567.4 Intron NP_001018861.3
XM_011521596.1 Intron XP_011519898.1
XM_011521597.1 Intron XP_011519899.1
XM_017022210.1 Intron XP_016877699.1
XM_017022211.1 Intron XP_016877700.1
XM_017022212.1 Intron XP_016877701.1
XM_017022213.1 Intron XP_016877702.1
XM_017022214.1 Intron XP_016877703.1
XM_017022215.1 Intron XP_016877704.1
XM_017022216.1 Intron XP_016877705.1
XM_017022217.1 Intron XP_016877706.1
XM_017022218.1 Intron XP_016877707.1

View Full Product Details