Product Details

SNP ID
rs4381490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52319177 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAATAACAGTAATTTGGGGAGA[C/T]AATAGTAACAACAGCAGCACTTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPTE2P2 PubMed Links

Gene Details

Gene
TPTE2P2
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 2
There are no transcripts associated with this gene.

View Full Product Details