Product Details

SNP ID
rs2134558
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30157085 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGATGGGTAATTTTTCAGCCCTC[A/G]AACCCCTCCTACCCTCGCTGCTTTT
Phenotype
MIM: 612963 MIM: 611940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCTN6 PubMed Links

Gene Details

Gene
DCTN6
Gene Name
dynactin subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006571.3 Intron NP_006562.1
Gene
MBOAT4
Gene Name
membrane bound O-acyltransferase domain containing 4
There are no transcripts associated with this gene.

View Full Product Details