Product Details

SNP ID
rs2838370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:43795072 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGGCCGGCTGTGTGCTGGGGCC[G/T]GCAGAGGTCACCACGGCCATGACTT
Phenotype
MIM: 610653
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RRP1 PubMed Links

Gene Details

Gene
RRP1
Gene Name
ribosomal RNA processing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003683.5 Intron NP_003674.1
XM_017028485.1 Intron XP_016883974.1
XM_017028486.1 Intron XP_016883975.1

View Full Product Details